Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009424.hg.1 |
TC05000679.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009424.hg.1 |
ENST00000274513.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009424.hg.1 |
ENST00000412661.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009424.hg.1 |
ENST00000420621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009424.hg.1 |
ENST00000433282.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009424.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000471980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000471980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000471980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000425402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000471980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000471980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000471980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
ENST00000510147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008730.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000274513.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000412661.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000433282.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
ENST00000462340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
NM_145282.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500160013.hg.1 |
TC0500008730.hg.1 |
Clariom D TC |