Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028269.hg.1 |
TC05002052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028269.hg.1 |
ENST00000274629.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028269.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028269.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028270.hg.1 |
TC05002052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028270.hg.1 |
ENST00000274629.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028270.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028270.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009401.hg.1 |
ENST00000518387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009401.hg.1 |
ENST00000518387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009401.hg.1 |
ENST00000518387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009401.hg.1 |
NR_109899.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009401.hg.1 |
NR_109899.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009401.hg.1 |
NR_109899.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009401.hg.1 |
ENST00000518387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009401.hg.1 |
ENST00000518387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009401.hg.1 |
ENST00000518387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009401.hg.1 |
NR_109899.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009401.hg.1 |
NR_109899.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009401.hg.1 |
NR_109899.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189048.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189048.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189048.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189048.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189048.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189048.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189048.hg.1 |
TC0500012791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500189050.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189050.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189050.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189050.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189050.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189050.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189050.hg.1 |
TC0500012791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
ENST00000521859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012791.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189051.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189051.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189051.hg.1 |
ENST00000274629.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189051.hg.1 |
NM_004137.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189051.hg.1 |
TC0500012791.hg.1 |
Clariom D TC |