Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011993.hg.1 |
ENST00000204517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011993.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011993.hg.1 |
TC16000827.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000574639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
ENST00000575672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009220.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000204517.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000572393.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000573476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000575300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
ENST00000575320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
NM_003223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600170520.hg.1 |
TC1600009220.hg.1 |
Clariom D TC |