Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015172.hg.1 |
ENST00000306357.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015172.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015172.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015172.hg.1 |
TC17001125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015173.hg.1 |
ENST00000306357.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015173.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015173.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015173.hg.1 |
TC17001125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009974.hg.1 |
ENST00000603161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000570987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000570987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000570987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573077.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573077.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000573881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000574775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000570583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000570987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000570987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000570987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573016.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573077.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573077.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000573881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000574775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009713.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175843.hg.1 |
TC1700009713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009974.hg.1 |
ENST00000603161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000306357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000574431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000575294.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
ENST00000575858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
NM_004853.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
NR_033656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175842.hg.1 |
TC1700009713.hg.1 |
Clariom D TC |