Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015059.2 | NP_055874.2 | 46 - 47 | 6359 | 82 | |
XM_005254708.4 | XP_005254765.1 | 49 - 50 | 6727 | 82 | ||
XM_005254710.4 | XP_005254767.1 | 50 - 51 | 6865 | 82 | ||
XM_005254711.4 | XP_005254768.1 | 49 - 50 | 7087 | 82 | ||
XM_005254712.4 | XP_005254769.1 | 49 - 50 | 6744 | 82 | ||
XM_005254713.4 | XP_005254770.1 | 49 - 50 | 7641 | 82 | ||
XM_005254714.3 | XP_005254771.1 | 48 - 49 | 6630 | 82 | ||
XM_005254715.2 | XP_005254772.1 | 48 - 49 | 7336 | 82 | ||
XM_006720717.3 | XP_006720780.1 | 49 - 50 | 7461 | 82 | ||
XM_017022665.1 | XP_016878154.1 | 49 - 50 | 7812 | 82 | ||
XM_017022666.1 | XP_016878155.1 | 49 - 50 | 6671 | 82 | ||
XM_017022667.1 | XP_016878156.1 | 47 - 48 | 6533 | 82 | ||
XM_017022668.1 | XP_016878157.1 | 49 - 50 | 7460 | 82 | ||
XM_017022669.1 | XP_016878158.1 | 48 - 49 | 7326 | 82 | ||
GenBank mRNA | AB002318.2 | - | 32 - 33 | 4533 | 82 | |
AB621808.1 | - | 27 - 28 | 3642 | 82 | ||
AF402000.1 | - | 46 - 47 | 6359 | 82 | ||
BX647654.1 | - | 17 - 18 | 4273 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |