Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000306821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000357731.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000434200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000467479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000467479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000467479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000467479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000467479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000474357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000474357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000478526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000478526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
ENST00000478526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014585.hg.1 |
NM_173808.2 |
Refseq |