Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000324446.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000324446.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000324446.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000454875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000454875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000454875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000324446.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000324446.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000324446.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000331787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000447937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000452584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000454875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000454875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000454875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
ENST00000610801.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_001543.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125734.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007143.hg.1 |
NR_125737.2 |
Refseq |