Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021151.hg.1 |
ENST00000369905.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021151.hg.1 |
ENST00000446605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021151.hg.1 |
ENST00000470322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021151.hg.1 |
ENST00000545684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021151.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021151.hg.1 |
TC10001623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179995.hg.1 |
TC1000011683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000369905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000470322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000480947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000481044.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000487599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
ENST00000494549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011683.hg.1 |
NM_005736.3 |
Refseq |