Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7968788 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7971242 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
222304_PM_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007983.hg.1 |
ENST00000503990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007983.hg.1 |
ENST00000503990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007983.hg.1 |
ENST00000503990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006927.hg.1 |
ENST00000592975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006850.hg.1 |
ENST00000514144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007097.hg.1 |
ENST00000400428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007097.hg.1 |
ENST00000400428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007097.hg.1 |
ENST00000400428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008728.hg.1 |
ENST00000419933.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008728.hg.1 |
ENST00000419933.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008728.hg.1 |
ENST00000419933.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011863.hg.1 |
ENST00000526052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011863.hg.1 |
ENST00000526052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011863.hg.1 |
ENST00000526052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007983.hg.1 |
ENST00000503990.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007983.hg.1 |
ENST00000503990.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007983.hg.1 |
ENST00000503990.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006850.hg.1 |
ENST00000514144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154615.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154615.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154615.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154615.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154615.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154615.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154615.hg.1 |
TC1200007655.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008728.hg.1 |
ENST00000419933.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008728.hg.1 |
ENST00000419933.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008728.hg.1 |
ENST00000419933.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154598.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154598.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154598.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154598.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154598.hg.1 |
TC1200007655.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200154597.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154597.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154597.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154597.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154597.hg.1 |
TC1200007655.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000442908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
ENST00000453589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011877.hg.1 |
NR_002822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007097.hg.1 |
ENST00000400428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007097.hg.1 |
ENST00000400428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007097.hg.1 |
ENST00000400428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011863.hg.1 |
ENST00000526052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011863.hg.1 |
ENST00000526052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011863.hg.1 |
ENST00000526052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006927.hg.1 |
ENST00000592975.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
216698_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
216698_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
222304_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
222304_PM_x_at |
- |
- |
Human Genome U133A 2.0 Array |
216698_x_at |
- |
- |
Human Genome U133A 2.0 Array |
216698_PM_x_at |
- |
- |
Human Genome U133A 2.0 Array |
222304_x_at |
- |
- |
Human Genome U133A 2.0 Array |
222304_PM_x_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw558724 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
222304_x_at5 |
- |
- |
Human Genome U133 Set |
216698_x_at |
- |
- |
Human Genome U133 Set |
222304_PM_x_at |
- |
- |
Human Genome U133 Set |
222304_x_at |
- |
- |
Human Genome U133 Set |
216698_PM_x_at |
- |
- |