Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134657.1 | NP_001128129.1 | 1 | 2690 | 99 | |
GenBank mRNA | AK128089.1 | - | 2 | 2553 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618146 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618154 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618148 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618142 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618159 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618161 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618157 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22758959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618136 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618133 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22758946 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618139 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22758957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22758948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22758961 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618156 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618158 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618135 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22758962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02618143 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 9368 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 210690 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3289917 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1843 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1842 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44688 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P36469512 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3289917 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3289917 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618156 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618142 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758946 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618158 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618154 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618142 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618158 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618156 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758946 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22758962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618154 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618161 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15493963 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618135 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618123 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15493952 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15493965 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618137 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618161 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618143 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618130 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618151 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618146 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618125 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15493951 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618141 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618150 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618148 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618156 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618144 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618154 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618133 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618127 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618158 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16423430 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8091045 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8091046 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8091044 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8091047 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8091043 | - | - |
Affymetrix Human Genome U219 Array | 11738358_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738358_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR03033117.hg.1 | TC03001830.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03033117.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03033117.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012591.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012591.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012591.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012591.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012591.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012591.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03033116.hg.1 | TC03001830.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03033116.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03033116.hg.1 | ENST00000502927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03033116.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03033118.hg.1 | TC03001830.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03033118.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03033118.hg.1 | ENST00000502927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03033118.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194159.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300194159.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194159.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300194159.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194159.hg.1 | TC0300012591.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300012591.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012591.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012591.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012591.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012591.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012591.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194157.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300194157.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194157.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300194157.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194157.hg.1 | TC0300012591.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300194158.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300194158.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194158.hg.1 | ENST00000413199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300194158.hg.1 | NM_001134657.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300194158.hg.1 | TC0300012591.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021663 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021663 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1731326 | AK128089.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1731326 | AK128089.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1731326 | AK128089.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1731326 | AK128089.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1731326 | AK128089.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1731326 | AK128089.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1731326 | AK128089.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1731326 | NM_001134657.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1731326 | AK128089.1 | human_transcriptome |