Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023904.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023904.hg.1 |
ENST00000538965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023904.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023904.hg.1 |
TC12001809.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023903.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023903.hg.1 |
ENST00000538965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023903.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023903.hg.1 |
TC12001809.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200189246.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189246.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189246.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189246.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189246.hg.1 |
TC1200011513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011513.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189245.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189245.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189245.hg.1 |
ENST00000378487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189245.hg.1 |
NM_001025232.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189245.hg.1 |
TC1200011513.hg.1 |
Clariom D TC |