Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010173.hg.1 |
TC04000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010173.hg.1 |
ENST00000438731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010173.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000508981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000508981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000508981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000510096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000510096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000510096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161259.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161259.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161259.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161259.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161259.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161259.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161259.hg.1 |
TC0400008913.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000438731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000508981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000508981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000508981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000510096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000510096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000510096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
ENST00000511965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008913.hg.1 |
NM_001080531.2 |
Refseq |