Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018472.hg.1 |
ENST00000261623.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018472.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018472.hg.1 |
TC16001339.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600187227.hg.1 |
TC1600011220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000261623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000561972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000562209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000563526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000565588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000566534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000567174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000568278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
ENST00000569359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011220.hg.1 |
NM_000101.3 |
Refseq |