Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003423.hg.1 |
TC21001068.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003423.hg.1 |
ENST00000470054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003423.hg.1 |
NM_182482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003423.hg.1 |
NM_182484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003422.hg.1 |
TC21001068.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100157724.hg.1 |
TC2100008486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100157724.hg.1 |
ENST00000470054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157724.hg.1 |
NM_182482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157724.hg.1 |
NM_182484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157724.hg.1 |
ENST00000470054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157724.hg.1 |
NM_182482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157724.hg.1 |
NM_182484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157725.hg.1 |
TC2100008486.hg.1 |
Clariom D TC |