Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000307236.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000381525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000428221.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000437573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000444832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000455198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000468775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000498701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000542100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_001278275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_001278276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_001278277.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_198256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_212540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NR_103490.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000307236.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000381525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000428221.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000437573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000444832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000455198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000468775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000498701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
ENST00000542100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_001278275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_001278276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_001278277.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_198256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NM_212540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
NR_103490.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185459.hg.1 |
TC0200011753.hg.1 |
Clariom D TC |