Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017021278.1 | XP_016876767.1 | 1 | 1077 | 100 | |
XM_017021281.1 | XP_016876770.1 | 1 | 1077 | 100 | ||
GenBank mRNA | BC041918.1 | - | 1 | 1354 | 100 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KTN1 antisense RNA 1 | KTN1-AS1 |
C14orf33, MYCLo-3 |
-2.7kb | Anti-sense | Chr. 14 - 55576157 - 55580092 on NCBI Build GRCh38 |
Downstream | ribosomal protein L13a pseudogene 3 | RPL13AP3 | RPL13A_11_1370 | 181.1kb | Sense | Chr. 14 - 55766245 - 55767717 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |