Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320674.1 | NP_001307603.1 | 11 | 5857 | 77 | |
NM_001320675.1 | NP_001307604.1 | 10 | 5616 | 77 | ||
NM_004330.3 | NP_004321.2 | 10 | 5821 | 77 | ||
GenBank mRNA | AK125533.1 | - | 1 | 2152 | 77 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | general transcription factor IIA subunit 2 | GTF2A2 |
HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS |
-1.6kb | Sense | Chr. 15 - 59638062 - 59657541 on NCBI Build GRCh38 |
Downstream | forkhead box B1 | FOXB1 |
FKH5, HFKH-5 |
342.4kb | Anti-sense | Chr. 15 - 60004222 - 60005943 on NCBI Build GRCh38 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |