Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1057 | LINC01057 | XLOC_000303 | Sense | Chr. 1 - 94657533 - 94820281 on NCBI Build GRCh38 | |
Downstream | microRNA 378g | MIR378G | 9.5kb | Sense | Chr. 1 - 94745860 - 94745900 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |