Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000368.hg.1 |
TC05000019.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000368.hg.1 |
ENST00000274137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000368.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000368.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000510329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
ENST00000510329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000510329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
ENST00000510329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006523.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144843.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144843.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144843.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144843.hg.1 |
ENST00000274137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144843.hg.1 |
ENST00000469176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144843.hg.1 |
NM_004553.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144843.hg.1 |
TC0500006523.hg.1 |
Clariom D TC |