Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015878.1 | NP_001015878.1 | 3 - 4 | 489 | 80 | |
NM_001015879.1 | NP_001015879.1 | 3 - 4 | 348 | 80 | ||
NM_003160.2 | NP_003151.2 | 3 - 4 | 353 | 80 | ||
GenBank mRNA | AB017332.1 | - | 3 - 4 | 494 | 80 | |
AF054621.1 | - | 3 - 4 | 494 | 80 | ||
AF059681.1 | - | 3 - 4 | 549 | 80 | ||
AK301886.1 | - | 2 - 3 | 929 | 80 | ||
AY661554.1 | - | 3 - 4 | 348 | 80 | ||
AY714054.1 | - | 3 - 4 | 291 | 80 | ||
BC075064.2 | - | 2 - 3 | 242 | 30915373 | 80 | |
EU668345.1 | - | 3 - 4 | 549 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons |