Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002711.3 | NP_002702.2 | 3 - 4 | 990 | 103 | |
XM_005250473.3 | XP_005250530.1 | 4 - 5 | 536 | 103 | ||
GenBank mRNA | AF024579.1 | - | 4 - 5 | 438 | 103 | |
BC126451.1 | - | 3 - 4 | 1028 | 8992167 | 103 | |
BC126453.1 | - | 3 - 4 | 1028 | 8992169 | 103 | |
X78578.1 | - | 3 - 4 | 957 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |