Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2AG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021064.4 | NP_066408.1 | 1 | 49 | 57 | |
GenBank mRNA | AK312124.1 | - | 1 | 49 | 57 | |
BC016677.1 | - | 1 | 34 | 4342191 | 57 | |
L19778.1 | - | 1 | 45 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |