Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.1 ST Array |
7923974 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057849.hg.1 |
TC01003775.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057849.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057849.hg.1 |
NR_029832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000385055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000385055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
NR_029518.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017137.hg.1 |
NR_029832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000385055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000385055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000415882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000487977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
NR_029518.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017137.hg.1 |
NR_029832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
NR_029832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000385231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
ENST00000608023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
NR_029832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229894.hg.1 |
TC0100017137.hg.1 |
Clariom D TC |