Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009773.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168152.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168152.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168152.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168152.hg.1 |
TC2000009773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168152.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168152.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168152.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168153.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168153.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168153.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168153.hg.1 |
TC2000009773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168153.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168153.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168153.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168154.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168154.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168154.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168154.hg.1 |
TC2000009773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168154.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168154.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168154.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168155.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168155.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168155.hg.1 |
NR_046225.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168155.hg.1 |
TC2000009773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168155.hg.1 |
ENST00000412500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168155.hg.1 |
NR_046224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168155.hg.1 |
NR_046225.1 |
Refseq |