Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL31RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242636.1 | NP_001229565.1 | 9 - 10 | 1431 | 88 | |
NM_001242637.1 | NP_001229566.1 | 9 - 10 | 1440 | 88 | ||
NM_001242638.1 | NP_001229567.1 | 11 - 12 | 1804 | 88 | ||
NM_001242639.1 | NP_001229568.1 | 10 - 11 | 1575 | 88 | ||
NM_001297570.1 | NP_001284499.1 | 9 - 10 | 1440 | 88 | ||
NM_001297572.1 | NP_001284501.1 | 9 - 10 | 1568 | 88 | ||
NM_139017.5 | NP_620586.3 | 9 - 10 | 1440 | 88 | ||
XM_011543142.2 | XP_011541444.1 | 9 - 10 | 1273 | 88 | ||
XM_011543143.2 | XP_011541445.1 | 9 - 10 | 1463 | 88 | ||
XM_011543144.2 | XP_011541446.1 | 8 - 9 | 1345 | 88 | ||
XM_011543145.2 | XP_011541447.1 | 6 - 7 | 914 | 88 | ||
GenBank mRNA | AF106913.1 | - | 9 - 10 | 1304 | 88 | |
AF486620.1 | - | 8 - 9 | 1152 | 88 | ||
AK297938.1 | - | 9 - 10 | 1552 | 88 | ||
AY358117.1 | - | 8 - 9 | 1152 | 88 | ||
AY358740.1 | - | 9 - 10 | 1440 | 88 | ||
AY499339.1 | - | 9 - 10 | 1313 | 88 | ||
AY499341.1 | - | 9 - 10 | 1313 | 88 | ||
AY499342.1 | - | 11 - 12 | 1687 | 88 | ||
BC110490.2 | - | 10 - 11 | 1367 | 40007489 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |