Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPALIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104296.1 | 2 - 3 | 570 | 69 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DNA nucleotidylexotransferase | DNTT | TDT | -12.8kb | Anti-sense | Chr. 10 - 96304328 - 96338564 on NCBI Build GRCh38 |
Downstream | tolloid like 2 | TLL2 | 9.3kb | Sense | Chr. 10 - 96364606 - 96513926 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |