Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000420226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000420226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000420226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000444939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000444939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000444939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000444939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
ENST00000452051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_046724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_121665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_121665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010010.hg.1 |
NR_121665.1 |
Refseq |