Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011513050.2 | XP_011511352.2 | 13 - 14 | 3009 | 104 | |
XM_017006939.1 | XP_016862428.1 | 14 - 15 | 3564 | 104 | ||
XM_017006940.1 | XP_016862429.1 | 14 - 15 | 3560 | 104 | ||
XM_017006941.1 | XP_016862430.1 | 14 - 15 | 3137 | 104 | ||
XM_017006942.1 | XP_016862431.1 | 14 - 15 | 3531 | 104 | ||
XM_017006943.1 | XP_016862432.1 | 14 - 15 | 3289 | 104 | ||
XM_017006944.1 | XP_016862433.1 | 13 - 14 | 3109 | 104 | ||
XM_017006945.1 | XP_016862434.1 | 13 - 14 | 2913 | 104 | ||
XM_017006946.1 | XP_016862435.1 | 12 - 13 | 2720 | 104 | ||
XM_017006947.1 | XP_016862436.1 | 11 - 12 | 2604 | 104 | ||
XM_017006948.1 | XP_016862437.1 | 13 - 14 | 2781 | 104 | ||
XM_017006949.1 | XP_016862438.1 | 12 - 13 | 2728 | 104 | ||
XM_017006950.1 | XP_016862439.1 | 12 - 13 | 8215 | 104 | ||
XM_017006951.1 | XP_016862440.1 | 12 - 13 | 8320 | 104 | ||
XM_017006952.1 | XP_016862441.1 | 11 - 12 | 2498 | 104 | ||
XM_017006953.1 | XP_016862442.1 | 11 - 12 | 2504 | 104 | ||
XM_017006954.1 | XP_016862443.1 | 5 - 6 | 1621 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Homo sapiens Probe spans exons |