Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569538.2 | XP_005569595.1 | 3 - 4 | 1762 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Probe spans exons |