Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566549.2 | XP_005566606.1 | 1 - 2 | 355 | 149 | |
XM_005566550.2 | XP_005566607.1 | 1 - 2 | 795 | 149 | ||
XM_005566551.2 | XP_005566608.1 | 1 - 2 | 884 | 149 | ||
XM_005566553.2 | XP_005566610.1 | 1 - 2 | 884 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |