Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTHLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570455.2 | XP_005570512.1 | 3 - 4 | 1169 | 74 | |
XM_015430568.1 | XP_015286054.1 | 3 - 4 | 859 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |