Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572407.2 | XP_005572464.1 | 4 - 5 | 474 | 69 | |
XM_005572408.2 | XP_005572465.1 | 4 - 5 | 521 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |