Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572407.2 | XP_005572464.1 | 1 - 2 | 92 | 102 | |
XM_005572408.2 | XP_005572465.1 | 1 - 2 | 93 | 102 | ||
XM_005572409.2 | XP_005572466.1 | 1 - 2 | 93 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |