Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577679.2 | XP_005577736.1 | 16 - 17 | 1919 | 110 | |
XM_005577680.2 | XP_005577737.1 | 16 - 17 | 1740 | 110 | ||
XM_015434639.1 | XP_015290125.1 | 17 - 18 | 1799 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |