Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015456902.1 | XP_015312388.1 | 4 - 5 | 447 | 143 | |
XM_015456903.1 | XP_015312389.1 | 4 - 5 | 415 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |