Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571319.2 | XP_005571376.1 | 1 - 2 | 1387 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |