Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551718.2 | XP_005551775.1 | 2 - 3 | 378 | 89 | |
XM_015448615.1 | XP_015304101.1 | 2 - 3 | 378 | 89 | ||
GenBank mRNA | AB172943.1 | - | 2 - 3 | 167 | 89 |