Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553103.2 | XP_005553160.1 | 5 - 6 | 860 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |