Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587227.2 | XP_005587284.1 | 2 - 3 | 285 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |