Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564063.2 | XP_005564120.1 | 4 - 5 | 1461 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |