Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569139.2 | XP_005569196.1 | 1 - 2 | 549 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |