Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589807.2 | XP_005589864.1 | 4 | 2190 | 127 | |
XM_005589810.2 | XP_005589867.1 | 3 | 1956 | 127 | ||
XM_015441771.1 | XP_015297257.1 | 2 | 1039 | 127 | ||
XM_015441772.1 | XP_015297258.1 | 2 | 963 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |