Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574252.2 | XP_005574309.1 | 4 | 2169 | 98 | |
XM_015432765.1 | XP_015288251.1 | 5 | 2211 | 98 | ||
XM_015432766.1 | XP_015288252.1 | 4 | 2136 | 98 | ||
XM_015432767.1 | XP_015288253.1 | 4 | 2133 | 98 | ||
XM_015432768.1 | XP_015288254.1 | 4 | 2480 | 98 | ||
XM_015432769.1 | XP_015288255.1 | 2 | 3518 | 98 | ||
XM_015432770.1 | XP_015288256.1 | 2 | 3448 | 98 | ||
XM_015432771.1 | XP_015288257.1 | 2 | 3415 | 98 | ||
XM_015432772.1 | XP_015288258.1 | 2 | 1860 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |