Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102123270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554812.2 | XP_005554869.1 | 2 | 178 | 78 | |
GenBank mRNA | AB169342.1 | - | 2 | 120 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Both primers and probe map within a single exon |