Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546151.2 | XP_005546208.1 | 1 | 1422 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |