Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589597.2 | XP_005589654.1 | 2 | 1112 | 96 | |
XM_005589598.2 | XP_005589655.2 | 2 | 1183 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |