Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283720.1 | NP_001270649.1 | 5 - 6 | 936 | 76 | |
XM_005575637.2 | XP_005575694.1 | 5 - 6 | 1086 | 76 | ||
XM_015433427.1 | XP_015288913.1 | 5 - 6 | 974 | 76 | ||
GenBank mRNA | AB168990.1 | - | 5 - 6 | 936 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |