Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285355.1 | NP_001272284.1 | 3 - 4 | 425 | 96 | |
XM_015447283.1 | XP_015302769.1 | 2 - 3 | 369 | 96 | ||
XM_015447284.1 | XP_015302770.1 | 2 - 3 | 1611 | 96 | ||
XM_015447285.1 | XP_015302771.1 | 2 - 3 | 732 | 96 | ||
XM_015447286.1 | XP_015302772.1 | 3 - 4 | 433 | 96 | ||
XM_015447287.1 | XP_015302773.1 | 2 - 3 | 370 | 96 | ||
XM_015447288.1 | XP_015302774.1 | 2 - 3 | 368 | 96 | ||
XM_015447289.1 | XP_015302775.1 | 2 - 3 | 369 | 96 | ||
XM_015447290.1 | XP_015302776.1 | 2 - 3 | 431 | 96 | ||
XM_015447292.1 | XP_015302778.1 | 2 - 3 | 369 | 96 | ||
GenBank mRNA | AB168194.1 | - | 3 - 4 | 425 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |