Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285245.1 | NP_001272174.1 | 5 - 6 | 1054 | 84 | |
XM_005560075.2 | XP_005560132.1 | 5 - 6 | 1536 | 84 | ||
XM_005560076.2 | XP_005560133.1 | 6 - 7 | 1274 | 84 | ||
XM_015452452.1 | XP_015307938.1 | 6 - 7 | 1290 | 84 | ||
XM_015452453.1 | XP_015307939.1 | 6 - 7 | 1215 | 84 | ||
GenBank mRNA | AB168338.1 | - | 5 - 6 | 1054 | 84 |