Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG10L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590097.2 | XP_005590154.1 | 9 - 10 | 4029 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |